Product Details

SNP ID
rs201192475
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:6416772 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGACCAGCCCACACTTGTGAGTGGG[A/G]ATGGAGAAGGTCATCTCCCCGCCAG
Phenotype
MIM: 603445 MIM: 610822
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
KHSRP PubMed Links

Gene Details

Gene
KHSRP
Gene Name
KH-type splicing regulatory protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003685.2 1258 Silent Mutation ATC,ATT I431I NP_003676.2
XM_005259668.4 1258 Silent Mutation ATC,ATT I431I XP_005259725.1
XM_011528395.2 1258 Silent Mutation ATC,ATT I387I XP_011526697.1
XM_017027408.1 1258 Silent Mutation ATC,ATT I387I XP_016882897.1
XM_017027409.1 1258 Silent Mutation ATC,ATT I387I XP_016882898.1
XM_017027410.1 1258 Silent Mutation ATC,ATT I287I XP_016882899.1
Gene
LOC390877
Gene Name
adenylate kinase isoenzyme 1-like
There are no transcripts associated with this gene.

Gene
MIR3940
Gene Name
microRNA 3940
There are no transcripts associated with this gene.

Gene
SLC25A41
Gene Name
solute carrier family 25 member 41
There are no transcripts associated with this gene.

View Full Product Details