Product Details

SNP ID
rs201645628
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:45350980 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATGAAGAGAGCCATGTCACTCAACA[C/T]ACTGAACGTGGATGCTCCAAGGGCT
Phenotype
MIM: 126340 MIM: 601334
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ERCC2 PubMed Links

Gene Details

Gene
ERCC2
Gene Name
ERCC excision repair 2, TFIIH core complex helicase subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000400.3 2961 Intron NP_000391.1
NM_001130867.1 2961 Intron NP_001124339.1
XM_011526611.2 2961 UTR 3 XP_011524913.1
XM_017026467.1 2961 UTR 3 XP_016881956.1
Gene
KLC3
Gene Name
kinesin light chain 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_177417.2 2961 Missense Mutation ACA,ATA T469I NP_803136.2

View Full Product Details