Product Details
- SNP ID
-
rs202032381
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.19:48837486 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GATCTGGGGGGAGGGGTCTCCTGGC[A/G]CGTGGGGTCCGAAGGAGGCAGCGGC
- Phenotype
-
MIM: 612832
MIM: 607769
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
HSD17B14
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs34460869] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- HSD17B14
- Gene Name
- hydroxysteroid 17-beta dehydrogenase 14
- Gene
- PLEKHA4
- Gene Name
- pleckstrin homology domain containing A4
View Full Product Details