Product Details

SNP ID
rs202204190
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:47801837 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCATTCACAGAGCCACCCTCCTCTT[A/G]GGGCTGAGACCCTGAGTGTTTTTTG
Phenotype
MIM: 611166
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TPRX1 PubMed Links

Gene Details

Gene
TPRX1
Gene Name
tetrapeptide repeat homeobox 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_198479.2 1245 Nonsense Mutation CAA,TAA Q392* NP_940881.2

View Full Product Details