Product Details

SNP ID
rs202237271
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:43718957 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACCTAAAGCAGGTAGACTTCAGCCG[C/G]TATGACTTCTTCCTGCTGGTCTCCC
Phenotype
MIM: 613176
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
IRGC PubMed Links

Gene Details

Gene
IRGC
Gene Name
immunity related GTPase cinema
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_019612.3 580 Silent Mutation CGC,CGG R133R NP_062558.1
XM_005259058.3 580 Silent Mutation CGC,CGG R170R XP_005259115.1
XM_006723283.3 580 Silent Mutation CGC,CGG R133R XP_006723346.1
XM_011527118.2 580 Silent Mutation CGC,CGG R133R XP_011525420.1
Gene
SMG9
Gene Name
SMG9, nonsense mediated mRNA decay factor
There are no transcripts associated with this gene.

View Full Product Details