Product Details

SNP ID
rs202101656
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:43652286 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCCTCGGCAGTCAATGAGGAAAGT[C/G]TCTTCAGAGGAGCATCCATGGGTGC
Phenotype
MIM: 173391
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
PLAUR PubMed Links

Gene Details

Gene
PLAUR
Gene Name
plasminogen activator, urokinase receptor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001005376.2 741 Missense Mutation GAC,GAG D231E NP_001005376.1
NM_001005377.2 741 Missense Mutation GAC,GAG D186E NP_001005377.1
NM_001301037.1 741 Intron NP_001287966.1
NM_002659.3 741 Missense Mutation GAC,GAG D231E NP_002650.1
XM_005258990.4 741 Missense Mutation GAC,GAG D231E XP_005259047.1
XM_011527027.2 741 Missense Mutation GAC,GAG D183E XP_011525329.1
XM_011527028.2 741 Missense Mutation ACA,AGA T238R XP_011525330.1
XM_011527029.1 741 Missense Mutation ACA,AGA T238R XP_011525331.1
XM_011527030.2 741 Intron XP_011525332.1
XM_011527031.2 741 Intron XP_011525333.1
XM_017026872.1 741 Missense Mutation GAC,GAG D226E XP_016882361.1
XM_017026873.1 741 Missense Mutation GAC,GAG D107E XP_016882362.1

View Full Product Details