Product Details

SNP ID
rs202206725
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:36182612 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAACTACGAATAAAGGCCATTCCAC[A/G]CTCCCTGCATTCATAGGGTTTCTCA
Phenotype
MIM: 614275
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ZNF565 PubMed Links

Gene Details

Gene
ZNF565
Gene Name
zinc finger protein 565
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001042474.2 2159 Missense Mutation CGT,TGT R452C NP_001035939.1
NM_152477.4 2159 Missense Mutation CGT,TGT R452C NP_689690.3
XM_011526512.2 2159 Missense Mutation CGT,TGT R452C XP_011524814.1
XM_011526514.2 2159 Missense Mutation CGT,TGT R452C XP_011524816.1
XM_017026341.1 2159 Missense Mutation CGT,TGT R452C XP_016881830.1
XM_017026342.1 2159 Missense Mutation CGT,TGT R387C XP_016881831.1
XM_017026343.1 2159 Missense Mutation CGT,TGT R387C XP_016881832.1

View Full Product Details