Product Details

SNP ID
rs201028602
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:48404983 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGTGCTCAGTGCCCAGCTCCGCAGT[A/G]TCAGCGCGCAGATCCGCCTGCTCTT
Phenotype
MIM: 602717
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GRIN2D PubMed Links

Gene Details

Gene
GRIN2D
Gene Name
glutamate ionotropic receptor NMDA type subunit 2D
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000836.2 803 Missense Mutation ATC,GTC I239V NP_000827.2
XM_011526872.1 803 Missense Mutation ATC,GTC I239V XP_011525174.1

View Full Product Details