Product Details

SNP ID
rs201095947
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:43607652 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCCGGGCATGCAGCATGAAGTGCC[C/T]GTGCAGCTCCCCCAGGTTGTCGAAG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SRRM5 PubMed Links

Gene Details

Gene
SRRM5
Gene Name
serine/arginine repetitive matrix 5
There are no transcripts associated with this gene.

Gene
ZNF428
Gene Name
zinc finger protein 428
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_182498.3 967 Missense Mutation AGG,GGG R178G NP_872304.2
Gene
ZNF576
Gene Name
zinc finger protein 576
There are no transcripts associated with this gene.

View Full Product Details