Product Details

SNP ID
rs201339514
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:9653392 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAGGACTTCTCTCCTTTATGAGTTT[C/T]TGCATGTGCAGAAAGTTGAGAAAAA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ZNF562 PubMed Links

Gene Details

Gene
ZNF562
Gene Name
zinc finger protein 562
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001130031.1 1190 Missense Mutation AAA,GAA K280E NP_001123503.1
NM_001130032.1 1190 Missense Mutation AAA,GAA K280E NP_001123504.1
NM_001300885.1 1190 Missense Mutation AAA,GAA K279E NP_001287814.1
NM_017656.3 1190 Missense Mutation AAA,GAA K208E NP_060126.2
XM_005259941.3 1190 Missense Mutation AAA,GAA K280E XP_005259998.1
XM_005259944.3 1190 Missense Mutation AAA,GAA K243E XP_005260001.1
XM_005259951.3 1190 Missense Mutation AAA,GAA K164E XP_005260008.1
XM_005259952.3 1190 Missense Mutation AAA,GAA K137E XP_005260009.1
XM_011528100.2 1190 Missense Mutation AAA,GAA K211E XP_011526402.1
XM_011528102.2 1190 Missense Mutation AAA,GAA K165E XP_011526404.1
XM_017026898.1 1190 Missense Mutation AAA,GAA K279E XP_016882387.1
XM_017026899.1 1190 Missense Mutation AAA,GAA K137E XP_016882388.1

View Full Product Details