Product Details

SNP ID
rs201409131
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:55132926 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCGGGGCGGCATCCTCACTTCCAGC[A/G]GCCTCCAACGCGGCCCTTCCCTGCC
Phenotype
MIM: 191041
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MIR7975 PubMed Links

Gene Details

Gene
MIR7975
Gene Name
microRNA 7975
There are no transcripts associated with this gene.

Gene
TNNT1
Gene Name
troponin T1, slow skeletal type
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001126132.2 1045 Missense Mutation CGC,TGC R260C NP_001119604.1
NM_001126133.2 1045 Missense Mutation CGC,TGC R249C NP_001119605.1
NM_001291774.1 1045 Missense Mutation CGC,TGC R249C NP_001278703.1
NM_003283.5 1045 Missense Mutation CGC,TGC R276C NP_003274.3
XM_011527246.2 1045 Missense Mutation CGC,TGC R256C XP_011525548.1
XM_017027186.1 1045 Missense Mutation CGC,TGC R260C XP_016882675.1
XM_017027187.1 1045 Missense Mutation CGC,TGC R256C XP_016882676.1

View Full Product Details