Product Details

SNP ID
rs202104062
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:56003724 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAAGATCCTCTTTTAAGTCTTGTCA[C/T]TCTTTCCACAGGTCCTACTTGCTCT
Phenotype
MIM: 609658
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
NLRP5 PubMed Links

Gene Details

Gene
NLRP5
Gene Name
NLR family pyrin domain containing 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_153447.4 71 Missense Mutation ACT,ATT T24I NP_703148.4

View Full Product Details