Product Details

SNP ID
rs202208907
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:9102657 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCAAGCTGCAAAATATATCAGGATG[A/T]TATTGATGAGGGTGTCTGAACAGGT
Phenotype
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
OR1M1 PubMed Links

Gene Details

Gene
OR1M1
Gene Name
olfactory receptor family 1 subfamily M member 1
There are no transcripts associated with this gene.

Gene
OR7G2
Gene Name
olfactory receptor family 7 subfamily G member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001005193.1 650 Missense Mutation AAC,ATC N217I NP_001005193.1

View Full Product Details