Product Details

SNP ID
rs200480165
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:49497292 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGAGAAGCTCCCGCGGTACTACAA[A/G]AACATCGGTCTGGGCTTCAAGACAC
Phenotype
MIM: 611114 MIM: 180471
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MIR150 PubMed Links

Gene Details

Gene
MIR150
Gene Name
microRNA 150
There are no transcripts associated with this gene.

Gene
RPL13A
Gene Name
ribosomal protein L13a
There are no transcripts associated with this gene.

Gene
RPS11
Gene Name
ribosomal protein S11
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001015.4 Intron NP_001006.1
Gene
SNORD32A
Gene Name
small nucleolar RNA, C/D box 32A
There are no transcripts associated with this gene.

Gene
SNORD33
Gene Name
small nucleolar RNA, C/D box 33
There are no transcripts associated with this gene.

Gene
SNORD34
Gene Name
small nucleolar RNA, C/D box 34
There are no transcripts associated with this gene.

Gene
SNORD35A
Gene Name
small nucleolar RNA, C/D box 35A
There are no transcripts associated with this gene.

Gene
SNORD35B
Gene Name
small nucleolar RNA, C/D box 35B
There are no transcripts associated with this gene.

View Full Product Details