Product Details

SNP ID
hCV190486762
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:42005823 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGTGGTATTCGTTCATGGTGGACTC[A/G]AGCAGGAAGGCGTAGCGGGAGTTGA
Phenotype
MIM: 600283
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GRIK5 PubMed Links

Gene Details

Gene
GRIK5
Gene Name
glutamate ionotropic receptor kainate type subunit 5
There are no transcripts associated with this gene.

View Full Product Details