Product Details

SNP ID
rs202179153
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:9186263 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTCATGACATCCCTCCTCCATATTT[C/G]TCTGATGATGCATCTAATCTTCTGT
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
OR7D2 PubMed Links

Gene Details

Gene
OR7D2
Gene Name
olfactory receptor family 7 subfamily D member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_175883.2 670 Missense Mutation TCT,TGT S161C NP_787079.1

View Full Product Details