Product Details

SNP ID
rs201201215
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:57193875 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTACTAATTCTGTTTGACTTTCCAG[A/G]TGTCTGTGACCTTTGATGATGTGGC
Phenotype
MIM: 604668
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ZNF264 PubMed Links

Gene Details

Gene
ZNF264
Gene Name
zinc finger protein 264
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003417.4 448 Missense Mutation ATG,GTG M12V NP_003408.1
XM_011527522.2 448 Missense Mutation ATG,GTG M12V XP_011525824.1

View Full Product Details