Product Details

SNP ID
rs201309448
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:57926182 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAAGGACTTCTTTCTGTGTGAACTC[C/T]CTGATGTCGAAGGAGAGAAGAGCTT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ZNF417 PubMed Links

Gene Details

Gene
ZNF417
Gene Name
zinc finger protein 417
There are no transcripts associated with this gene.

Gene
ZNF418
Gene Name
zinc finger protein 418
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001317027.1 2305 Missense Mutation AGA,GGA R688G NP_001303956.1
NM_001317028.1 2305 Missense Mutation AGA,GGA R668G NP_001303957.1
NM_001317029.1 2305 Missense Mutation AGA,GGA R667G NP_001303958.1
NM_001317030.1 2305 Missense Mutation AGA,GGA R582G NP_001303959.1
NM_133460.2 2305 Missense Mutation AGA,GGA R667G NP_597717.1
XM_017026304.1 2305 Missense Mutation AGA,GGA R688G XP_016881793.1
XM_017026305.1 2305 Missense Mutation AGA,GGA R688G XP_016881794.1
XM_017026306.1 2305 Missense Mutation AGA,GGA R687G XP_016881795.1
XM_017026307.1 2305 Missense Mutation AGA,GGA R677G XP_016881796.1
XM_017026308.1 2305 Missense Mutation AGA,GGA R677G XP_016881797.1
XM_017026309.1 2305 Missense Mutation AGA,GGA R668G XP_016881798.1
XM_017026310.1 2305 Missense Mutation AGA,GGA R668G XP_016881799.1
XM_017026311.1 2305 Missense Mutation AGA,GGA R668G XP_016881800.1
XM_017026312.1 2305 Missense Mutation AGA,GGA R667G XP_016881801.1
XM_017026313.1 2305 Missense Mutation AGA,GGA R667G XP_016881802.1
XM_017026314.1 2305 Missense Mutation AGA,GGA R634G XP_016881803.1

View Full Product Details