Product Details

SNP ID
rs201931758
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:2717288 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGCGCTTGCCGTCGATGTTGAGGCT[C/T]ATGTTCCGGCGCGTCTCCAGCGTCA
Phenotype
MIM: 607862
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DIRAS1 PubMed Links

Gene Details

Gene
DIRAS1
Gene Name
DIRAS family GTPase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_145173.3 842 Missense Mutation ATA,ATG I173M NP_660156.1
XM_011527713.2 842 Missense Mutation ATA,ATG I207M XP_011526015.1

View Full Product Details