Product Details

SNP ID
rs201379859
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.19:14472635 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCAGGCGCTCGGTGTTAGGCCCAG[A/C]CCCGCGGGGCCGCCCTTGGCTCCGG
Phenotype
MIM: 605072 MIM: 601032 MIM: 176802
Polymorphism
A/C, Transversion Substitution
Allele Nomenclature
Literature Links
GIPC1 PubMed Links

Gene Details

Gene
GIPC1
Gene Name
GIPC PDZ domain containing family member 1
There are no transcripts associated with this gene.

Gene
PKN1
Gene Name
protein kinase N1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002741.3 1252 Intron NP_002732.3
NM_213560.1 1252 Intron NP_998725.1
XM_011528128.1 1252 Intron XP_011526430.1
Gene
PTGER1
Gene Name
prostaglandin E receptor 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000955.2 1252 Silent Mutation GGG,GGT G378G NP_000946.2

View Full Product Details