Product Details

SNP ID
rs201793120
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:19120834 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCGTCAGGTACTGCAAGCTCACCA[C/G]GGTCACATAGCAGTAGACTCGGACC
Phenotype
MIM: 610823
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
SLC25A42 PubMed Links

Gene Details

Gene
SLC25A42
Gene Name
solute carrier family 25 member 42
There are no transcripts associated with this gene.

Gene
TMEM161A
Gene Name
transmembrane protein 161A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001256766.1 1086 Missense Mutation CTG,GTG L270V NP_001243695.1
NM_017814.2 1086 Missense Mutation CTG,GTG L373V NP_060284.1
XM_011528117.2 1086 Missense Mutation CTG,GTG L171V XP_011526419.1
XM_017026916.1 1086 Missense Mutation CTG,GTG L348V XP_016882405.1

View Full Product Details