Product Details

SNP ID
rs200967897
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:48630963 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTTCTCCAGGAAGGCCGCCCGCAC[C/T]GATATCTGGTTCTCCTTGAGCCGCC
Phenotype
MIM: 604644 MIM: 124097 MIM: 607092
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CA11 PubMed Links

Gene Details

Gene
CA11
Gene Name
carbonic anhydrase 11
There are no transcripts associated with this gene.

Gene
DBP
Gene Name
D-box binding PAR bZIP transcription factor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001352.4 1106 Silent Mutation TCA,TCG S284S NP_001343.2
XM_017026388.1 1106 Silent Mutation TCA,TCG S141S XP_016881877.1
Gene
SEC1P
Gene Name
secretory blood group 1, pseudogene
There are no transcripts associated with this gene.

Gene
SPHK2
Gene Name
sphingosine kinase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001204158.2 1106 Intron NP_001191087.1
NM_001204159.2 1106 Intron NP_001191088.1
NM_001204160.2 1106 Intron NP_001191089.1
NM_001243876.1 1106 Intron NP_001230805.1
NM_020126.4 1106 Intron NP_064511.2
XM_006723292.1 1106 Intron XP_006723355.1
XM_011527133.1 1106 Intron XP_011525435.1
XM_011527134.1 1106 Intron XP_011525436.1
XM_017027008.1 1106 Intron XP_016882497.1
XM_017027009.1 1106 Intron XP_016882498.1
XM_017027010.1 1106 Intron XP_016882499.1

View Full Product Details