Product Details

SNP ID
rs201104772
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:5691326 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGCCCCTTTCCCCCCTAGCGTCTGA[A/C]CAAACACACCAAGTTCGTGCGGGAC
Phenotype
MIM: 605490
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
HSD11B1L PubMed Links

Gene Details

Gene
HSD11B1L
Gene Name
hydroxysteroid 11-beta dehydrogenase 1 like
There are no transcripts associated with this gene.

Gene
LONP1
Gene Name
lon peptidase 1, mitochondrial
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001276479.1 Intron NP_001263408.1
NM_001276480.1 Intron NP_001263409.1
NM_004793.3 Intron NP_004784.2
XM_011528441.2 Intron XP_011526743.1
Gene
RPL36
Gene Name
ribosomal protein L36
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015414.3 Intron NP_056229.2
NM_033643.2 Intron NP_378669.1

View Full Product Details