Product Details
- SNP ID
-
rs201278961
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.19:5831514 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CAGGTGAACCAAGCCGCTATGCCGC[A/G]TGTCTGGTACCTGGATTCCTCCTGC
- Phenotype
-
MIM: 136836
MIM: 602018
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
FUT6
PubMed Links
Gene Details
- Gene
- FUT6
- Gene Name
- fucosyltransferase 6
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_000150.2 |
2388 |
Missense Mutation |
CGC,TGC |
R352C |
NP_000141.1 |
NM_001040701.1 |
2388 |
Missense Mutation |
CGC,TGC |
R352C |
NP_001035791.1 |
XM_005259526.4 |
2388 |
Missense Mutation |
CGC,TGC |
R352C |
XP_005259583.1 |
XM_005259527.3 |
2388 |
Intron |
|
|
XP_005259584.1 |
XM_011527868.2 |
2388 |
Missense Mutation |
CGC,TGC |
R352C |
XP_011526170.1 |
XM_011527869.2 |
2388 |
Missense Mutation |
CGC,TGC |
R352C |
XP_011526171.1 |
XM_011527870.2 |
2388 |
Missense Mutation |
CGC,TGC |
R352C |
XP_011526172.1 |
XM_011527872.2 |
2388 |
Missense Mutation |
CGC,TGC |
R352C |
XP_011526174.1 |
XM_011527873.2 |
2388 |
Missense Mutation |
CGC,TGC |
R352C |
XP_011526175.1 |
XM_011527874.2 |
2388 |
Missense Mutation |
CGC,TGC |
R352C |
XP_011526176.1 |
XM_011527875.2 |
2388 |
Missense Mutation |
CGC,TGC |
R352C |
XP_011526177.1 |
XM_011527876.2 |
2388 |
Missense Mutation |
CGC,TGC |
R352C |
XP_011526178.1 |
XM_011527878.2 |
2388 |
Missense Mutation |
CGC,TGC |
R352C |
XP_011526180.1 |
XM_011527879.2 |
2388 |
Missense Mutation |
CGC,TGC |
R352C |
XP_011526181.1 |
- Gene
- NRTN
- Gene Name
- neurturin
There are no transcripts associated with this gene.
View Full Product Details