Product Details

SNP ID
rs201658217
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:17240680 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCAGCACGTACTCACCCTCCTTCC[G/T]CATGCCCACCCGGAAGCACTTCTTG
Phenotype
MIM: 132880 MIM: 611810
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
NR2F6 PubMed Links

Gene Details

Gene
NR2F6
Gene Name
nuclear receptor subfamily 2 group F member 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005234.3 486 Silent Mutation AGG,CGG R122R NP_005225.2
Gene
USHBP1
Gene Name
USH1 protein network component harmonin binding protein 1
There are no transcripts associated with this gene.

View Full Product Details