Product Details

SNP ID
rs200662076
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:8324691 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAGCAGCAGCCGCACGGTGTCCAGG[C/T]GCCCATACTCACTGGCACACATCAG
Phenotype
MIM: 614611 MIM: 602139 MIM: 603685
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
KANK3 PubMed Links

Gene Details

Gene
KANK3
Gene Name
KN motif and ankyrin repeat domains 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_198471.2 2288 Missense Mutation CAC,CGC H741R NP_940873.2
XM_006722718.3 2288 Missense Mutation CAC,CGC H741R XP_006722781.1
XM_011527884.2 2288 Missense Mutation CAC,CGC H741R XP_011526186.1
XM_017026563.1 2288 Missense Mutation CAC,CGC H792R XP_016882052.1
Gene
NDUFA7
Gene Name
NADH:ubiquinone oxidoreductase subunit A7
There are no transcripts associated with this gene.

Gene
RPS28
Gene Name
ribosomal protein S28
There are no transcripts associated with this gene.

View Full Product Details