Product Details

SNP ID
rs200796739
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:17283427 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGATTACAGCTCAGACGCCTCTTT[C/T]GTCACAGCGGTTGAGGTCTCTGGAG
Phenotype
MIM: 612766
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ABHD8 PubMed Links

Gene Details

Gene
ABHD8
Gene Name
abhydrolase domain containing 8
There are no transcripts associated with this gene.

Gene
ANKLE1
Gene Name
ankyrin repeat and LEM domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001278443.1 934 Silent Mutation TTC,TTT F264F NP_001265372.1
NM_001278444.1 934 Silent Mutation TTC,TTT F275F NP_001265373.1
NM_001278445.1 934 Silent Mutation TTC,TTT F243F NP_001265374.1
NM_152363.5 934 Silent Mutation TTC,TTT F275F NP_689576.5
Gene
BABAM1
Gene Name
BRISC and BRCA1 A complex member 1
There are no transcripts associated with this gene.

View Full Product Details