Product Details

SNP ID
rs200833450
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:8399909 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCGTGGGCAAGAGCAACCTGCTGTC[A/G]CGCTTCACCCGCAACGAGTTCAACC
Phenotype
MIM: 604198
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
RAB11B PubMed Links

Gene Details

Gene
RAB11B
Gene Name
RAB11B, member RAS oncogene family
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004218.3 183 Silent Mutation TCA,TCG S29S NP_004209.2
Gene
RAB11B-AS1
Gene Name
RAB11B antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details