Product Details

SNP ID
rs200900605
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:50819254 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCACATAAGACAGCACTCTGACGGC[A/G]ACAGAAGGCTTATTGGGGGTGCCAC
Phenotype
MIM: 147910 MIM: 610601
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
KLK1 PubMed Links

Gene Details

Gene
KLK1
Gene Name
kallikrein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002257.3 1664 Silent Mutation NP_002248.1
XM_011526942.1 1664 Silent Mutation XP_011525244.1
Gene
KLK15
Gene Name
kallikrein related peptidase 15
There are no transcripts associated with this gene.

Gene
MGC45922
Gene Name
uncharacterized LOC284365
There are no transcripts associated with this gene.

View Full Product Details