Product Details

SNP ID
rs200901862
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:35545758 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGGCATCATGACCCTGTTTCACTTC[C/G]GGAACTGCTTCGCTCTTGCCTACTT
Phenotype
MIM: 137216 MIM: 609169 MIM: 613585
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
ATP4A PubMed Links

Gene Details

Gene
ATP4A
Gene Name
ATPase H+/K+ transporting alpha subunit
There are no transcripts associated with this gene.

Gene
GAPDHS
Gene Name
glyceraldehyde-3-phosphate dehydrogenase, spermatogenic
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014364.4 159 Intron NP_055179.1
XM_017026579.1 159 Intron XP_016882068.1
Gene
TMEM147
Gene Name
transmembrane protein 147
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001242597.1 159 UTR 5 NP_001229526.1
NM_001242598.1 159 Missense Mutation CGG,GGG R7G NP_001229527.1
NM_032635.3 159 Missense Mutation CGG,GGG R7G NP_116024.1
Gene
TMEM147-AS1
Gene Name
TMEM147 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details