Product Details

SNP ID
rs201487991
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:57978528 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGTACAATAAGGGATGAACTCTCAT[G/T]AAATGCTTTCCCACATTCATTACAC
Phenotype
MIM: 613905
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
C19orf18 PubMed Links

Gene Details

Gene
C19orf18
Gene Name
chromosome 19 open reading frame 18
There are no transcripts associated with this gene.

Gene
ZNF606
Gene Name
zinc finger protein 606
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_025027.3 2630 Missense Mutation AAT,CAT N718H NP_079303.2
XM_005259276.1 2630 Missense Mutation AAT,CAT N718H XP_005259333.1

View Full Product Details