Product Details
- SNP ID
-
rs201762230
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.19:56414048 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CTGCTCAGAGGAATTTGTACAGGAA[A/C]GTGATGTTGGAGAACTACAGGAGCT
- Phenotype
-
- Polymorphism
- A/C, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
ZNF583
PubMed Links
Gene Details
- Gene
- ZNF583
- Gene Name
- zinc finger protein 583
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001159860.1 |
2100 |
Missense Mutation |
AAA,AAC |
K33N |
NP_001153332.1 |
NM_001159861.1 |
2100 |
Missense Mutation |
AAA,AAC |
K33N |
NP_001153333.1 |
NM_152478.2 |
2100 |
Missense Mutation |
AAA,AAC |
K33N |
NP_689691.2 |
XM_005258558.1 |
2100 |
Missense Mutation |
AAA,AAC |
K34N |
XP_005258615.1 |
XM_011526517.2 |
2100 |
Missense Mutation |
AAA,AAC |
K33N |
XP_011524819.1 |
XM_017026346.1 |
2100 |
Missense Mutation |
AAA,AAC |
K33N |
XP_016881835.1 |
XM_017026347.1 |
2100 |
Missense Mutation |
AAA,AAC |
K33N |
XP_016881836.1 |
XM_017026348.1 |
2100 |
Missense Mutation |
AAA,AAC |
K33N |
XP_016881837.1 |
XM_017026349.1 |
2100 |
Missense Mutation |
AAA,AAC |
K33N |
XP_016881838.1 |
XM_017026350.1 |
2100 |
Missense Mutation |
AAA,AAC |
K33N |
XP_016881839.1 |
XM_017026351.1 |
2100 |
UTR 5 |
|
|
XP_016881840.1 |
XM_017026352.1 |
2100 |
UTR 5 |
|
|
XP_016881841.1 |
XM_017026353.1 |
2100 |
UTR 5 |
|
|
XP_016881842.1 |
View Full Product Details