Product Details

SNP ID
rs201904181
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:48498116 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCCCATTCACCAGCACCTTCTCCC[C/T]GGCCTCTGGGGACCTCAGCTCCCCA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LMTK3 PubMed Links

Gene Details

Gene
LMTK3
Gene Name
lemur tyrosine kinase 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001080434.1 3371 Missense Mutation AGG,GGG R1014G NP_001073903.1
XM_011526411.2 3371 Missense Mutation AGG,GGG R1011G XP_011524713.1
XM_011526412.2 3371 Missense Mutation AGG,GGG R1000G XP_011524714.1

View Full Product Details