Product Details
- SNP ID
-
rs202009324
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.19:12015764 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ATAAAAGGCTTTCCCACATATCTTA[C/G]ATTTATGAGATATCTCTCCAGTGTG
- Phenotype
-
- Polymorphism
- C/G, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
LOC101928464
PubMed Links
Gene Details
- Gene
- LOC101928464
- Gene Name
- uncharacterized LOC101928464
There are no transcripts associated with this gene.
- Gene
- ZNF433
- Gene Name
- zinc finger protein 433
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001080411.2 |
1343 |
Missense Mutation |
TCT,TGT |
S368C |
NP_001073880.1 |
NM_001308346.1 |
1343 |
Missense Mutation |
TCT,TGT |
S367C |
NP_001295275.1 |
NM_001308348.1 |
1343 |
Missense Mutation |
TCT,TGT |
S365C |
NP_001295277.1 |
NM_001308351.1 |
1343 |
Missense Mutation |
TCT,TGT |
S333C |
NP_001295280.1 |
NM_001308355.1 |
1343 |
Missense Mutation |
TCT,TGT |
S333C |
NP_001295284.1 |
NM_001308357.1 |
1343 |
Missense Mutation |
TCT,TGT |
S333C |
NP_001295286.1 |
XM_006722661.3 |
1343 |
Missense Mutation |
TCT,TGT |
S333C |
XP_006722724.1 |
XM_011527725.2 |
1343 |
Missense Mutation |
TCT,TGT |
S379C |
XP_011526027.1 |
XM_011527728.2 |
1343 |
Missense Mutation |
TCT,TGT |
S367C |
XP_011526030.1 |
XM_011527729.2 |
1343 |
Missense Mutation |
TCT,TGT |
S367C |
XP_011526031.1 |
XM_017026413.1 |
1343 |
Intron |
|
|
XP_016881902.1 |
View Full Product Details