Product Details

SNP ID
rs200834388
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:6678402 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTGATGGTCTGCTCAATGGCCATGA[G/T]GTACTCGTCAAAGTCATTGGACAGC
Phenotype
MIM: 120700 MIM: 604520
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
C3 PubMed Links

Gene Details

Gene
C3
Gene Name
complement component 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000064.3 4777 Missense Mutation ATC,CTC I1562L NP_000055.2
Gene
TNFSF14
Gene Name
tumor necrosis factor superfamily member 14
There are no transcripts associated with this gene.

View Full Product Details