Product Details

SNP ID
rs200869444
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:49118884 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTCTGCGTGGGAGTCGCGGGTGGGG[A/G]TGCCGCGTGCAGTTCCCGAGGCGGG
Phenotype
MIM: 612331 MIM: 603144
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
C19orf73 PubMed Links

Gene Details

Gene
C19orf73
Gene Name
chromosome 19 open reading frame 73
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018111.2 257 Missense Mutation CCC,TCC P47S NP_060581.2
Gene
LIN7B
Gene Name
lin-7 homolog B, crumbs cell polarity complex component
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001308419.1 257 Intron NP_001295348.1
NM_022165.2 257 Intron NP_071448.1
XM_006723323.3 257 Intron XP_006723386.1
XM_017027131.1 257 Intron XP_016882620.1
Gene
PPFIA3
Gene Name
PTPRF interacting protein alpha 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003660.3 257 Intron NP_003651.1
XM_017027407.1 257 Intron XP_016882896.1

View Full Product Details