Product Details
- SNP ID
-
rs201106981
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.19:37738652 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ACGACTGAAGGCCTTCCCACATTCT[G/T]TACATTCATAAGGTTTTCCACCAGT
- Phenotype
-
- Polymorphism
- G/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
ZNF573
PubMed Links
Gene Details
- Gene
- ZNF573
- Gene Name
- zinc finger protein 573
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001172689.1 |
1774 |
Missense Mutation |
AAA,ACA |
K525T |
NP_001166160.1 |
NM_001172690.1 |
1774 |
Missense Mutation |
AAA,ACA |
K613T |
NP_001166161.1 |
NM_001172691.1 |
1774 |
Missense Mutation |
AAA,ACA |
K611T |
NP_001166162.1 |
NM_001172692.1 |
1774 |
Missense Mutation |
AAA,ACA |
K525T |
NP_001166163.1 |
NM_152360.3 |
1774 |
Missense Mutation |
AAA,ACA |
K555T |
NP_689573.3 |
XM_011526446.2 |
1774 |
Missense Mutation |
AAA,ACA |
K628T |
XP_011524748.1 |
XM_017026275.1 |
1774 |
Missense Mutation |
AAA,ACA |
K628T |
XP_016881764.1 |
XM_017026276.1 |
1774 |
Missense Mutation |
AAA,ACA |
K628T |
XP_016881765.1 |
XM_017026277.1 |
1774 |
Missense Mutation |
AAA,ACA |
K626T |
XP_016881766.1 |
XM_017026278.1 |
1774 |
Missense Mutation |
AAA,ACA |
K557T |
XP_016881767.1 |
XM_017026279.1 |
1774 |
Missense Mutation |
AAA,ACA |
K557T |
XP_016881768.1 |
XM_017026280.1 |
1774 |
Missense Mutation |
AAA,ACA |
K525T |
XP_016881769.1 |
XM_017026281.1 |
1774 |
Missense Mutation |
AAA,ACA |
K525T |
XP_016881770.1 |
View Full Product Details