Product Details

SNP ID
rs201107314
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:49689906 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCGCCCAAGGGGACCTGGACACTGC[A/G]GCCAGGTGAGAAACCCGGGTGGCAG
Phenotype
MIM: 608846 MIM: 602950
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ADM5 PubMed Links

Gene Details

Gene
ADM5
Gene Name
adrenomedullin 5 (putative)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001101340.1 1222 Silent Mutation GCA,GCG A23A NP_001094810.1
Gene
CPT1C
Gene Name
carnitine palmitoyltransferase 1C
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001136052.2 1222 Intron NP_001129524.1
NM_001199752.2 1222 Intron NP_001186681.1
NM_001199753.1 1222 Intron NP_001186682.1
NM_152359.2 1222 Intron NP_689572.1
XM_005258505.2 1222 Intron XP_005258562.1
XM_005258506.4 1222 Intron XP_005258563.1
XM_006723009.3 1222 Intron XP_006723072.1
XM_011526438.2 1222 Intron XP_011524740.1
XM_011526439.2 1222 Intron XP_011524741.1
XM_011526440.2 1222 Intron XP_011524742.1
XM_017026265.1 1222 Intron XP_016881754.1
XM_017026266.1 1222 Intron XP_016881755.1
XM_017026267.1 1222 Intron XP_016881756.1
XM_017026268.1 1222 Intron XP_016881757.1
XM_017026269.1 1222 Intron XP_016881758.1
XM_017026270.1 1222 Intron XP_016881759.1
XM_017026271.1 1222 Intron XP_016881760.1
XM_017026272.1 1222 Intron XP_016881761.1
XM_017026273.1 1222 Intron XP_016881762.1
Gene
MIR5088
Gene Name
microRNA 5088
There are no transcripts associated with this gene.

Gene
PRMT1
Gene Name
protein arginine methyltransferase 1
There are no transcripts associated with this gene.

View Full Product Details