Product Details

SNP ID
rs201559740
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:36182625 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGCCATTCCACACTCCCTGCATTC[A/G]TAGGGTTTCTCACAAGTGTGAATTC
Phenotype
MIM: 614275
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ZNF565 PubMed Links

Gene Details

Gene
ZNF565
Gene Name
zinc finger protein 565
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001042474.2 2146 Silent Mutation TAC,TAT Y447Y NP_001035939.1
NM_152477.4 2146 Silent Mutation TAC,TAT Y447Y NP_689690.3
XM_011526512.2 2146 Silent Mutation TAC,TAT Y447Y XP_011524814.1
XM_011526514.2 2146 Silent Mutation TAC,TAT Y447Y XP_011524816.1
XM_017026341.1 2146 Silent Mutation TAC,TAT Y447Y XP_016881830.1
XM_017026342.1 2146 Silent Mutation TAC,TAT Y382Y XP_016881831.1
XM_017026343.1 2146 Silent Mutation TAC,TAT Y382Y XP_016881832.1

View Full Product Details