Product Details

SNP ID
rs201695916
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:9828611 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCCAGCACGGATGATACCATCGGG[A/G]ACCTTAAGAAGCTGATTGCAGCCCA
Phenotype
MIM: 609079 MIM: 601052 MIM: 606849
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FBXL12 PubMed Links

Gene Details

Gene
FBXL12
Gene Name
F-box and leucine rich repeat protein 12
There are no transcripts associated with this gene.

Gene
PIN1
Gene Name
peptidylprolyl cis/trans isomerase, NIMA-interacting 1
There are no transcripts associated with this gene.

Gene
UBL5
Gene Name
ubiquitin like 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001048241.2 Intron NP_001041706.1
NM_024292.3 Intron NP_077268.1

View Full Product Details