Product Details
- SNP ID
-
rs201696002
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.19:9185998 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CAACCTGTCCTGGGTTGACATCTGT[C/T]TCAGCACTTGCATCGTCCCCAAGAT
- Phenotype
-
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
OR7D2
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs56246934] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- OR7D2
- Gene Name
- olfactory receptor family 7 subfamily D member 2
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_175883.2 |
405 |
Missense Mutation |
CTC,TTC |
L73F |
NP_787079.1 |
View Full Product Details