Product Details

SNP ID
rs202187420
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:12689390 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACCTTACCCTATTGAGCCCATTGTC[A/G]TGCCTTCGGGTGCAAATGGTCCTTG
Phenotype
MIM: 600944 MIM: 609074 MIM: 603002
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DHPS PubMed Links

Gene Details

Gene
DHPS
Gene Name
deoxyhypusine synthase
There are no transcripts associated with this gene.

Gene
FBXW9
Gene Name
F-box and WD repeat domain containing 9
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032301.2 1463 Silent Mutation CAC,CAT H428H NP_115677.2
XM_005260096.4 1463 Silent Mutation CAC,CAT H265H XP_005260153.1
Gene
LOC105372280
Gene Name
uncharacterized LOC105372280
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001316692.1 1463 Intron NP_001303621.1
Gene
TNPO2
Gene Name
transportin 2
There are no transcripts associated with this gene.

View Full Product Details