Product Details

SNP ID
rs202187613
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:20007128 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAGTCCATTATAAATTTCTTTTTGA[C/T]CCTTACACTCACCCACATTTTCCCC
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ZNF682 PubMed Links

Gene Details

Gene
ZNF682
Gene Name
zinc finger protein 682
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001077349.1 601 Intron NP_001070817.1
NM_033196.2 601 Intron NP_149973.1
XM_017027455.1 601 Missense Mutation GAT,GGT D131G XP_016882944.1
XM_017027456.1 601 Intron XP_016882945.1

View Full Product Details