Product Details

SNP ID
rs201007765
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:34759419 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATGTCGAATAAAAACAGAGTGGTGT[A/G]TAAAAGCCTTTCCACATTCACTGCA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ZNF599 PubMed Links

Gene Details

Gene
ZNF599
Gene Name
zinc finger protein 599
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001007248.2 1770 Missense Mutation ACA,ATA T461I NP_001007249.1
XM_017026369.1 1770 Missense Mutation ACA,ATA T424I XP_016881858.1

View Full Product Details