Product Details

SNP ID
rs201076191
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:4446183 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCGTACACCGCCCCCAGCCGCTCCC[A/G]AGCGTAGAAAGTGCCTGGGGAGTGG
Phenotype
MIM: 601246 MIM: 611946
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CHAF1A PubMed Links

Gene Details

Gene
CHAF1A
Gene Name
chromatin assembly factor 1 subunit A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005483.2 1594 Intron NP_005474.2
XM_011527605.2 1594 Intron XP_011525907.1
XM_011527606.2 1594 Intron XP_011525908.1
XM_011527607.2 1594 Intron XP_011525909.1
Gene
MIR4746
Gene Name
microRNA 4746
There are no transcripts associated with this gene.

Gene
UBXN6
Gene Name
UBX domain protein 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001171091.1 1594 Silent Mutation NP_001164562.1
NM_025241.2 1594 Silent Mutation NP_079517.1
XM_017027325.1 1594 Missense Mutation XP_016882814.1

View Full Product Details