Product Details

SNP ID
rs201144869
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:58356567 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTGCCGCACTGGGCGCACGCATGAG[C/G]CTTCTCGCCCGTGTGCACGCGCCGG
Phenotype
MIM: 138670
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
A1BG PubMed Links

Gene Details

Gene
A1BG
Gene Name
alpha-1-B glycoprotein
There are no transcripts associated with this gene.

Gene
A1BG-AS1
Gene Name
A1BG antisense RNA 1
There are no transcripts associated with this gene.

Gene
LOC100419840
Gene Name
zinc finger protein 446 pseudogene
There are no transcripts associated with this gene.

Gene
LOC105372483
Gene Name
uncharacterized LOC105372483
There are no transcripts associated with this gene.

Gene
ZNF497
Gene Name
zinc finger protein 497
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001207009.1 1255 Missense Mutation CCT,GCT P357A NP_001193938.1
NM_198458.2 1255 Missense Mutation CCT,GCT P357A NP_940860.2

View Full Product Details