Product Details

SNP ID
rs201595252
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:19266513 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACCTGGCCGATGCCTCCAGCAAACA[A/C]CAAGGCCCAGTCTGGAAGCCAGGAG
Phenotype
MIM: 607992 MIM: 606563
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
HAPLN4 PubMed Links

Gene Details

Gene
HAPLN4
Gene Name
hyaluronan and proteoglycan link protein 4
There are no transcripts associated with this gene.

Gene
SUGP1
Gene Name
SURP and G-patch domain containing 1
There are no transcripts associated with this gene.

Gene
TM6SF2
Gene Name
transmembrane 6 superfamily member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001001524.2 951 Missense Mutation GTG,TTG V301L NP_001001524.2

View Full Product Details