Product Details

SNP ID
rs201495282
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:36183129 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAGCCTTGCCACAGTCTTTACATAC[A/G]TAGGGTTTCTCGCCTGTGTGAGTTC
Phenotype
MIM: 614275
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ZNF565 PubMed Links

Gene Details

Gene
ZNF565
Gene Name
zinc finger protein 565
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001042474.2 1642 Silent Mutation TAC,TAT Y279Y NP_001035939.1
NM_152477.4 1642 Silent Mutation TAC,TAT Y279Y NP_689690.3
XM_011526512.2 1642 Silent Mutation TAC,TAT Y279Y XP_011524814.1
XM_011526514.2 1642 Silent Mutation TAC,TAT Y279Y XP_011524816.1
XM_017026341.1 1642 Silent Mutation TAC,TAT Y279Y XP_016881830.1
XM_017026342.1 1642 Silent Mutation TAC,TAT Y214Y XP_016881831.1
XM_017026343.1 1642 Silent Mutation TAC,TAT Y214Y XP_016881832.1

View Full Product Details