Product Details

SNP ID
rs201804406
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:19679326 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGCGTGTGTGGGAAAGTCTTCCTC[C/T]GTCATTCATTCCTGGACAGGCACAT
Phenotype
MIM: 603983
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ZNF101 PubMed Links

Gene Details

Gene
ZNF101
Gene Name
zinc finger protein 101
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001300949.1 Intron NP_001287878.1
NM_033204.3 Intron NP_149981.2

View Full Product Details