Product Details

SNP ID
rs201980724
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:463882 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCGGGCAGCCTGTGCCCCGCAGGCC[A/G]GGAGGGCGTGGTGGCGGCCATGGTG
Phenotype
MIM: 605217
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ODF3L2 PubMed Links

Gene Details

Gene
ODF3L2
Gene Name
outer dense fiber of sperm tails 3 like 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_182577.2 638 Missense Mutation CGG,TGG R278W NP_872383.1
XM_005259545.4 638 Missense Mutation CGG,TGG R170W XP_005259602.1
XM_017026671.1 638 Missense Mutation CGG,TGG R170W XP_016882160.1
Gene
SHC2
Gene Name
SHC adaptor protein 2
There are no transcripts associated with this gene.

View Full Product Details